4-(2-Aminothiazol-4-yl)phenol 97%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Amino-4-thiazolyl)phenol

A653768-10G A653768-1G A653768-250MG
77267-192EA 6.18 USD
77267-192 77267-194 77267-196 77267-198
4-(2-Aminothiazol-4-yl)phenol 97%
4-(2-Aminothiazol-4-yl)phenol

Precaución: for Research Use Only

Formula: C₉H₈N₂OS
MDL Number: MFCD01624171
CAS Number: 57634-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR