(E)-3-(3-Bromo-4-fluorophenyl)acrylic acid 98%

Supplier: Ambeed
Warning

A653607-25G A653607-5G
77267-188EA 76.22 USD
77267-188 77267-190
(E)-3-(3-Bromo-4-fluorophenyl)acrylic acid 98%
(E)-3-(3-Bromo-4-fluorophenyl)acrylic acid

Precaución: for Research Use Only

Formula: C₉H₆BrFO₂
MW: 245.04 g/mol
MDL Number: MFCD00143095
CAS Number: 370867-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR