2-(2-Fluorophenyl)-1H-imidazo[4,5-c]pyridine 99%
Supplier: Ambeed
|
Warning
|
A653291-100MG
A653291-250MG
77267-184EA
9
USD
77267-184
77267-186
2-(2-Fluorophenyl)-1H-imidazo[4,5-c]pyridine 99%
2-(2-Fluorophenyl)-1H-imidazo[4,5-c]pyridine
Precaución: for Research Use Only
Formula:
C₁₂H₈FN₃ |
CAS Number:
89075-43-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...