9,3':6',9''-Tercarbazole 98%

Supplier: Ambeed
Warning

A652761-1G A652761-250MG
77243-054EA 112 USD
77243-054 77243-056
9,3':6',9''-Tercarbazole 98%
9,3':6',9''-Tercarbazole

Precaución: for Research Use Only

Formula: C₃₆H₂₃N₃
MDL Number: MFCD29472452
CAS Number: 606129-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR