Benzyl tert-butyl (2-oxopropane-1,3-diyl)dicarbamate 97%

Supplier: Ambeed
Warning

A652683-100MG A652683-250MG
77267-174EA 62.83 USD
77267-174 77267-176
Benzyl tert-butyl (2-oxopropane-1,3-diyl)dicarbamate 97%
Benzyl tert-butyl (2-oxopropane-1,3-diyl)dicarbamate

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₂O₅
MW: 322.35 g/mol
MDL Number: MFCD30607319
CAS Number: 877317-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR