Azomethine H sodium salt 98%

Supplier: Ambeed
Warning

Synonyms: Sodium hydrogen 4-hydroxy-5-(salicylideneamino)naphthalene-2,7-disulphonate, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid 6-sodium salt, Azomethine H, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid monosodium salt, Azomethine-H monosodium salt

A652117-1G A652117-25G A652117-5G
77243-042EA 29 USD
77243-042 77243-044 77243-046
Azomethine H sodium salt 98%
Azomethine H sodium salt

Precaución: for Research Use Only

Formula: C₁₇H₁₂NNaO₈S₂
MW: 445.41 g/mol
MDL Number: MFCD00042050
CAS Number: 5941-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR