cis-2,6-Dimethylpiperazine 98%

Supplier: Ambeed
Danger

A651921-10G A651921-25G A651921-5G
77254-718EA 12 USD
77254-718 77254-720 77254-722
cis-2,6-Dimethylpiperazine 98%
cis-2,6-Dimethylpiperazine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
MDL Number: MFCD07772435
CAS Number: 21655-48-1
UN: 1325
ADR: 4.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR