2-Ethylanthracene 98+%

Supplier: Ambeed
Danger

A651841-100MG A651841-1G A651841-250MG A651841-5G
77267-164EA 28 USD
77267-164 77267-166 77267-168 77267-170
2-Ethylanthracene 98+%
2-Ethylanthracene

Precaución: for Research Use Only

Formula: C₁₆H₁₄
MDL Number: MFCD00003584
CAS Number: 52251-71-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR