3-(Trimethylsilyl)-2-oxazolidinone 98%

Supplier: Ambeed
Danger

Synonyms: 3-(Trimethylsilyl)oxazolidin-2-one

A651670-1G A651670-5G
77293-040EA 8 USD
77293-040 77293-042
3-(Trimethylsilyl)-2-oxazolidinone 98%
3-(Trimethylsilyl)-2-oxazolidinone

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂Si
MW: 159.26 g/mol
MDL Number: MFCD00010197
CAS Number: 43112-38-5
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR