1-Chloro-2,2-dimethylpropane 98%

Supplier: Ambeed
Danger

Synonyms: Chloro-1-diméthyl-2,2-propane, neo-Pentylchloride

A651604-1G A651604-5G A651604-25G
77243-038EA 11 USD
77243-038 77243-040 77551-972
1-Chloro-2,2-dimethylpropane 98%
1-Chloro-2,2-dimethylpropane

Precaución: for Research Use Only

Formula: C₅H₁₁Cl
MW: 106.6 g/mol
Boiling Pt: 84
Melting Pt: -20
MDL Number: MFCD00000941
CAS Number: 753-89-9
UN: 1993
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR