3-Methylisatoic anhydride 98%

Supplier: Ambeed
Warning

Synonyms: 8-Methyl-1H-benzo[d][1,3]oxazine-2,4-dione

A651541-100MG A651541-1G A651541-250MG A651541-25G A651541-5G
77267-154EA 11.33 USD
77267-154 77267-156 77267-158 77267-160 77267-162
3-Methylisatoic anhydride 98%
3-Methylisatoic anhydride

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MW: 177.1568 g/mol
MDL Number: MFCD03426399
CAS Number: 66176-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR