2,3-Dichloro-α,α,α-trifluorotoluene 98%
Supplier: Ambeed
|
Warning
|
A651506-1000G
A651506-100G
A651506-10G
A651506-1G
A651506-25G
A651506-500G
A651506-5G
77254-702EA
425.39
USD
77254-702
77254-704
77254-706
77254-708
77254-710
77254-712
77254-714
2,3-Dichloro-α,α,α-trifluorotoluene 98%
2,3-Dichloro-α,α,α-trifluorotoluene
Precaución: for Research Use Only
Formula:
C₇H₃Cl₂F₃ MW: 215 g/mol |
MDL Number:
MFCD00069115 CAS Number: 54773-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...