4-Chloro-6-iodo-7-methoxyquinazoline 97%

Supplier: Ambeed
Warning

A650934-100MG A650934-1G A650934-250MG
77254-690EA 27 USD
77254-690 77254-692 77254-694
4-Chloro-6-iodo-7-methoxyquinazoline 97%
4-Chloro-6-iodo-7-methoxyquinazoline

Precaución: for Research Use Only

Formula: C₉H₆ClIN₂O
MW: 320.51 g/mol
MDL Number: MFCD12973651
CAS Number: 1172851-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR