4-Boc(2-aminomethyl)morpholine 97%

Supplier: Ambeed
Danger

A649620-1G A649620-5G
77267-148EA 53 USD
77267-148 77267-150
4-Boc(2-aminomethyl)morpholine 97%
4-Boc(2-aminomethyl)morpholine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₃
MDL Number: MFCD06410633
CAS Number: 140645-53-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR