GNE-140 98%

Supplier: Ambeed
Warning

Synonyms: GNE-140 racemate

A649509-10MG A649509-1MG A649509-5MG A649509-50MG
77267-142EA 576.8 USD
77267-142 77267-144 77267-146 77749-335
GNE-140 98%
GNE-140

Precaución: for Research Use Only

Formula: C₂₅H₂₃ClN₂O₃S₂
MW: 499.04 g/mol
MDL Number: MFCD30489754
CAS Number: 1802977-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR