(3S,4S,5R,6R)-6-((Benzoyloxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetrabenzoate 97%

Supplier: Ambeed
Warning

A649446-100G A649446-25G A649446-5G
77243-020EA 165.83 USD
77243-020 77243-022 77243-024
(3S,4S,5R,6R)-6-((Benzoyloxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetrabenzoate 97%
(3S,4S,5R,6R)-6-((Benzoyloxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetrabenzoate

Precaución: for Research Use Only

Formula: C₄₁H₃₂O₁₁
MW: 700.68 g/mol
MDL Number: MFCD02683389
CAS Number: 96996-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR