5-Methylfuran-2(5H)-one 95%

Supplier: Ambeed
Warning

A649352-100MG A649352-1G A649352-250MG
77243-018EA 46.35 USD
77243-018 77573-084 77747-334
5-Methylfuran-2(5H)-one 95%
5-Methylfuran-2(5H)-one

Precaución: for Research Use Only

Formula: C₅H₆O₂
MW: 98.09 g/mol
MDL Number: MFCD01725808
CAS Number: 591-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR