2-[(4-Bromophenoxy)methyl]oxirane 95%

Supplier: Ambeed
Danger

A649268-100MG A649268-1G A649268-250MG A649268-5G
77254-672EA 13 USD
77254-672 77254-674 77254-676 77254-678
2-[(4-Bromophenoxy)methyl]oxirane 95%
2-[(4-Bromophenoxy)methyl]oxirane

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MDL Number: MFCD00022341
CAS Number: 2212-06-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR