3-(Morpholinomethyl)benzoic acid 97%

Supplier: Ambeed
Warning

A648964-1G A648964-250MG A648964-5G
77293-026EA 17 USD
77293-026 77293-028 77293-030
3-(Morpholinomethyl)benzoic acid 97%
3-(Morpholinomethyl)benzoic acid

Precaución: for Research Use Only

Formula: C12H15NO3
MDL Number: MFCD05841588
CAS Number: 67451-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR