2-Bromo-1,8-naphthyridine 95%

Supplier: Ambeed
Warning

A648943-100MG A648943-250MG
77267-140EA 46 USD
77267-140 77617-728
2-Bromo-1,8-naphthyridine 95%
2-Bromo-1,8-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₅BrN₂
MDL Number: MFCD00234385
CAS Number: 61323-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR