2,2,3,3-Tetramethyl-4,7,10-trioxa-3-siladodecan-12-ol 98%

Supplier: Ambeed
Warning

A648705-100MG A648705-1G A648705-250MG A648705-5G
77267-132EA 30.9 USD
77267-132 77267-134 77267-136 77267-138
2,2,3,3-Tetramethyl-4,7,10-trioxa-3-siladodecan-12-ol 98%
2,2,3,3-Tetramethyl-4,7,10-trioxa-3-siladodecan-12-ol

Precaución: for Research Use Only

Formula: C₁₂H₂₈O₄Si
MW: 264.43 g/mol
MDL Number: MFCD30471718
CAS Number: 201037-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR