Ethyl-3-(3-boronobenzamido)propionate 98%

Supplier: Ambeed
Warning

Synonyms: 3-[(3-Ethoxy-3-oxopropyl)carbamoyl]benzeneboronic acid, (3-((3-Ethoxy-3-oxopropyl)carbamoyl)phenyl)boronic acid

A648461-1G A648461-250MG A648461-5G
77280-524EA 161.71 USD
77280-524 77280-526 77280-528
Ethyl-3-(3-boronobenzamido)propionate 98%
Ethyl 3-(3-boronobenzamido)propionate

Precaución: for Research Use Only

Formula: C₁₂H₁₆BNO₅
MW: 265.07 g/mol
MDL Number: MFCD04115701
CAS Number: 850567-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR