2-Methyl-2-hexenoic acid 95%

Supplier: Ambeed
Danger

A648358-1G A648358-250MG A648358-5G
77267-126EA 44 USD
77267-126 77267-128 77267-130
2-Methyl-2-hexenoic acid 95%
2-Methyl-2-hexenoic acid

Precaución: for Research Use Only

Formula: C₇H₁₂O₂
MDL Number: MFCD00015855
CAS Number: 28897-58-7
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR