[1,3-Bis(2,6-di-i-propylphenyl)imidazol-2-ylidene][bis(trifluoromethanesulfonyl)imide]gold(I) 98%

Supplier: Ambeed
Warning

A647761-100MG A647761-250MG
77254-668EA 48 USD
77254-668 77254-670
[1,3-Bis(2,6-di-i-propylphenyl)imidazol-2-ylidene][bis(trifluoromethanesulfonyl)imide]gold(I) 98%
[1,3-Bis(2,6-di-i-propylphenyl)imidazol-2-ylidene][bis(trifluoromethanesulfonyl)imide]gold(I)

Precaución: for Research Use Only

Formula: C₂₉H₃₆AuF₆N₃O₄S₂
MW: 865.70 g/mol
MDL Number: MFCD22572651
CAS Number: 951776-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR