(14aS)-1,14-Bis(diphenylphosphino)-6,7,8,9-tetrahydrodibenzo[b,d][1,6]dioxecine 98%

Supplier: Ambeed
Warning

A647718-100MG A647718-1G A647718-250MG
77267-120EA 100 USD
77267-120 77267-122 77267-124
(14aS)-1,14-Bis(diphenylphosphino)-6,7,8,9-tetrahydrodibenzo[b,d][1,6]dioxecine 98%
(14aS)-1,14-Bis(diphenylphosphino)-6,7,8,9-tetrahydrodibenzo[b,d][1,6]dioxecine

Precaución: for Research Use Only

Formula: C₄₀H₃₄O₂P₂
CAS Number: 486429-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR