2-(2-((tert-Butyldimethylsilyl)oxy)ethoxy)ethanol 97%
Supplier: Ambeed
|
Warning
|
A647710-1G
A647710-25G
A647710-5G
A647710-500G
A647710-100G
77254-662EA
9.27
USD
77254-662
77254-664
77254-666
77747-331
77747-330
2-(2-((tert-Butyldimethylsilyl)oxy)ethoxy)ethanol 97%
2-(2-((tert-Butyldimethylsilyl)oxy)ethoxy)ethanol
Precaución: for Research Use Only
Formula:
C₁₀H₂₄O₃Si MW: 220.38 g/mol |
MDL Number:
MFCD27943432 CAS Number: 131326-39-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...