4-Oxo-4-(2-thienyl)butyric acid 98%

Supplier: Ambeed
Warning

A646248-10G A646248-1G A646248-250MG A646248-5G
77293-012EA 74 USD
77293-012 77293-014 77293-016 77293-018
4-Oxo-4-(2-thienyl)butyric acid 98%
4-Oxo-4-(2-thienyl)butyric acid

Precaución: for Research Use Only

Formula: C₈H₈O₃S
MDL Number: MFCD00022502
CAS Number: 4653-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR