1,3-Dichloro-4-fluoro-6-nitrobenzene 96%

Supplier: Ambeed
Warning

Synonyms: 1,5-Dichloro-2-fluoro-4-nitrobenzene, 2,4-Dichloro-5-fluoronitrobenzene

A646238-1G A646238-25G A646238-5G
77254-650EA 18.54 USD
77254-650 77254-652 77254-654
1,3-Dichloro-4-fluoro-6-nitrobenzene 96%
1,3-Dichloro-4-fluoro-6-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₂FNO₂
MW: 209.99 g/mol
MDL Number: MFCD00115003
CAS Number: 2105-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR