4-Methyl-1-naphthol 97%

Supplier: Ambeed
Warning

A645734-100MG A645734-1G A645734-250MG A645734-5G
77293-004EA 38 USD
77293-004 77293-006 77293-008 77293-010
4-Methyl-1-naphthol 97%
4-Methyl-1-naphthol

Precaución: for Research Use Only

Formula: C₁₁H₁₀O
MDL Number: MFCD00060344
CAS Number: 10240-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR