(E)-3-(3-Hydroxy-4-methoxyphenyl)acrylic acid 97%

Supplier: Ambeed
Warning

A645594-10G A645594-1G A645594-25G A645594-5G
77267-104EA 99.91 USD
77267-104 77267-106 77267-108 77267-110
(E)-3-(3-Hydroxy-4-methoxyphenyl)acrylic acid 97%
(E)-3-(3-Hydroxy-4-methoxyphenyl)acrylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.184 g/mol
MDL Number: MFCD00004391
CAS Number: 25522-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR