GDC-0853 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-(3'-(hydroxymethyl)-1-methyl-5-((5-(2-methyl-4-(oxetan-3-yl)piperazin-1-yl)pyridin-2-yl)amino)-6-oxo-1,6-dihydro-[3,4'-bipyridin]-2'-yl)-7,7-dimethyl-3,4,7,8-tetrahydro-2H-cyclopenta[4,5]pyrrolo[1,2-a]pyrazin-1(6H)-one, Fenebrutinib
A645470-10MG
A645470-1MG
A645470-25MG
A645470-2MG
A645470-5MG
77267-094EA
149.35
USD
77267-094
77267-096
77267-098
77267-100
77267-102
GDC-0853 98+%
GDC-0853
Precaución: for Research Use Only
Formula:
C₃₇H₄₄N₈O₄ MW: 664.81 g/mol |
CAS Number:
1434048-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...