Diethyl 4-((4-bromo-2-cyanophenyl)carbamoyl)benzylphosphonate 97%

Supplier: Ambeed
Danger

A644717-10MG A644717-1MG A644717-25MG A644717-50MG A644717-5MG
77254-638EA 55 USD
77254-638 77254-640 77254-642 77254-644 77254-646
Diethyl 4-((4-bromo-2-cyanophenyl)carbamoyl)benzylphosphonate 97%
Diethyl 4-((4-bromo-2-cyanophenyl)carbamoyl)benzylphosphonate

Precaución: for Research Use Only

Formula: C₁₉H₂₀BrN₂O₄P
MDL Number: MFCD00897807
CAS Number: 133208-93-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR