3-Amino-3-(2,4-dichlorophenyl)-propionic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Amino-3-(2,4-dichlorophenyl)propionic acid
A644659-1G
A644659-250MG
77280-506EA
18.54
USD
77280-506
77280-508
77280-510
3-Amino-3-(2,4-dichlorophenyl)-propionic acid 95%
3-Amino-3-(2,4-dichlorophenyl)-propionic acid
Precaución: for Research Use Only
Formula:
C₉H₉Cl₂NO₂ |
MDL Number:
MFCD02663185 CAS Number: 152606-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...