(2S,5R)-5-((Benzyloxy)amino)piperidine-2-carboxamide 97%

Supplier: Ambeed
Warning

A644536-1G A644536-250MG A644536-5G
77254-632EA 43.26 USD
77254-632 77254-634 77254-636
(2S,5R)-5-((Benzyloxy)amino)piperidine-2-carboxamide 97%
(2S,5R)-5-((Benzyloxy)amino)piperidine-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₃H₁₉N₃O₂
MW: 249.30 g/mol
MDL Number: MFCD29472345
CAS Number: 1416134-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR