Penta-O-acetyl-ɑ-D-glucopyranose 97%

Supplier: Ambeed
Warning

Synonyms: 1,2,3,4,6-Penta-O-acetyl-α-D-glycopyranose, ɑ-D-Glucose Pentaacetate, 1,2,3,4,6-Penta-O-Acetyl-A-D-Glycopyranose, α-D(+)-Glucose pentaacetate, (2R,3R,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate, Pentaacetyl-ɑ-D-glucose, α-D-Glucose pentaacetate

A644266-100G A644266-25G A644266-500G A644266-1000G
77280-500EA 11 USD
77280-500 77280-502 77280-504 77749-329
Penta-O-acetyl-ɑ-D-glucopyranose 97%
Penta-O-acetyl-alpha-D-glucopyranose

Precaución: for Research Use Only

Formula: C₁₆H₂₂O₁₁
MDL Number: MFCD00064071
CAS Number: 604-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR