(±)-1-Phenyl-2,2,2-trifluoroethanol 98%

Supplier: Ambeed
Warning

Synonyms: α-(Trifluoromethyl)benzyl alcohol, 2,2,2-Trifluoro-1-phenylethan-1-ol

A644131-1G A644131-25G A644131-5G
77243-004EA 25.75 USD
77243-004 77243-006 77243-008
(±)-1-Phenyl-2,2,2-trifluoroethanol 98%
(±)-1-Phenyl-2,2,2-trifluoroethanol

Precaución: for Research Use Only

Formula: C₆H₅CH(OH)CF₃
MW: 176.14 g/mol
MDL Number: MFCD00004498
CAS Number: 340-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR