3'-Chloro-5'-fluoroacetophenone 98%

Supplier: Ambeed
Warning

A644030-1G A644030-250MG A644030-5G
77280-492EA 18 USD
77280-492 77280-494 77280-496
3'-Chloro-5'-fluoroacetophenone 98%
3'-Chloro-5'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆ClFO
MDL Number: MFCD04115834
CAS Number: 842140-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR