9H-Dibenzo[a,c]carbazole 97%

Supplier: Ambeed
Warning

A643844-250MG A643844-5G
77242-996EA 17.51 USD
77242-996 77242-998
9H-Dibenzo[a,c]carbazole 97%
9H-Dibenzo[a,c]carbazole

Precaución: for Research Use Only

Formula: C₂₀H₁₃N
MW: 267.32 g/mol
CAS Number: 201-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR