Ruthenium, dichlorotetrakis(triphenylphosphine)- 98%

Supplier: Ambeed
Warning

A643836-1G A643836-250MG
77254-628EA 40 USD
77254-628 77254-630
Ruthenium, dichlorotetrakis(triphenylphosphine)- 98%
Ruthenium, dichlorotetrakis(triphenylphosphine)-

Precaución: for Research Use Only

Formula: C₇₂H₆₀Cl₂P₄Ru
MW: 1221.11 g/mol
MDL Number: MFCD09039275
CAS Number: 15555-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR