Tetrahydro-2H-pyran-2-ol 97%

Supplier: Ambeed
Warning

Synonyms: ~1-Valerolactol, 2-Hydroxytetrahydropyran

A643657-5G A643657-25G
77267-086EA 53.56 USD
77267-086 77551-952
Tetrahydro-2H-pyran-2-ol 97%
Tetrahydro-2H-pyran-2-ol

Precaución: for Research Use Only

Formula: C₅H₁₀O₂
MW: 102.133 g/mol
MDL Number: MFCD02683093
CAS Number: 694-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR