3-(N-Phenyl-N-methyl)aminoacrolein 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Methyl(phenyl)amino)acrylaldehyde

A643402-1G A643402-250MG A643402-5G
77292-982EA 111.24 USD
77292-982 77292-984 77292-986
3-(N-Phenyl-N-methyl)aminoacrolein 98%
3-(N-Phenyl-N-methyl)aminoacrolein

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
MDL Number: MFCD00190652
CAS Number: 14189-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR