Dorzagliatin 97%

Supplier: Ambeed
Warning

A642740-10MG A642740-25MG A642740-5MG A642740-1MG A642740-50MG
77254-622EA 67 USD
77254-622 77254-624 77254-626 77617-716 77747-324
Dorzagliatin 97%
Dorzagliatin

Precaución: for Research Use Only

Formula: C₂₂H₂₇ClN₄O₅
MDL Number: MFCD22376579
CAS Number: 1191995-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR