Manganese bis(trifluoromethanesulfonate) 98%

Supplier: Ambeed
Danger

A642659-100MG A642659-1G A642659-250MG A642659-25G A642659-5G
77267-068EA 5 USD
77267-068 77267-070 77267-072 77267-074 77267-076
Manganese bis(trifluoromethanesulfonate) 98%
Manganese bis(trifluoromethanesulfonate)

Precaución: for Research Use Only

Formula: 2(CF₃O₃S)·Mn
MDL Number: MFCD17015267
CAS Number: 55120-76-8
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR