4-(Pyrimidin-2-ylmethyl)-7-(4-(trifluoromethoxy)phenyl)-3,4-dihydrobenzo[f][1,4]oxazepin-5(2H)-one hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Eleclazine HCl
A642597-1MG
A642597-5MG
A642597-50MG
77267-064EA
15
USD
77267-064
77267-066
77749-319
4-(Pyrimidin-2-ylmethyl)-7-(4-(trifluoromethoxy)phenyl)-3,4-dihydrobenzo[f][1,4]oxazepin-5(2H)-one hydrochloride 98%
4-(Pyrimidin-2-ylmethyl)-7-(4-(trifluoromethoxy)phenyl)-3,4-dihydrobenzo[f][1,4]oxazepin-5(2H)-one hydrochloride
Precaución: for Research Use Only
Formula:
C₂₁H₁₇ClF₃N₃O₃ |
MDL Number:
MFCD30377215 CAS Number: 1448754-43-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...