2,3,4,6-Tetra-O-pivaloyl-α-D-glucopyranosyl bromide 98%

Supplier: Ambeed
Warning

Synonyms: 2,3,4,6-Tetra-O-pivaloyl-alpha-D-glucopyranosyl bromide, (2R,3R,4S,5R,6R)-2-Bromo-6-((pivaloyloxy)methyl)tetrahydro-2H-pyran-3,4,5-triyl tris(2,2-dimethylpropanoate)

A642508-100G A642508-25G A642508-5G
77254-616EA 91.67 USD
77254-616 77254-618 77747-323
2,3,4,6-Tetra-O-pivaloyl-α-D-glucopyranosyl bromide 98%
2,3,4,6-Tetra-O-pivaloyl-α-D-glucopyranosyl bromide

Precaución: for Research Use Only

Formula: C₂₆H₄₃BrO₉
MW: 579.518 g/mol
MDL Number: MFCD08275217
CAS Number: 81058-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR