2-(1H-Imidazol-5-yl)acetic acid 95%

Supplier: Ambeed
Warning

A642448-100MG A642448-1G A642448-250MG
77242-990EA 54.99 USD
77242-990 77242-992 77242-994
2-(1H-Imidazol-5-yl)acetic acid 95%
2-(1H-Imidazol-5-yl)acetic acid

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MDL Number: MFCD08668377
CAS Number: 645-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR