3-Amino-5-bromobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A641970-100G
A641970-10G
A641970-1G
A641970-25G
A641970-5G
77280-482EA
159
USD
77280-482
77280-484
77280-486
77280-488
77280-490
3-Amino-5-bromobenzoic acid 97%
3-Amino-5-bromobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₆BrNO₂ |
MDL Number:
MFCD00227745 CAS Number: 42237-85-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...