3-Hydroxy-3-methylbutanone 98%

Supplier: Ambeed
Danger

Synonyms: 3-Hydroxy-3-methyl-2-butanone, 3-Hydroxy-3-methylbutan-2-one

A641513-1G A641513-25G A641513-5G
77280-476EA 18 USD
77280-476 77280-478 77280-480
3-Hydroxy-3-methylbutanone 98%
3-Hydroxy-3-methylbutanone

Precaución: for Research Use Only

Formula: C₅H₁₀O₂
MDL Number: MFCD00004460
CAS Number: 115-22-0
UN: 1224
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR