(S)-3,3'-Bis(4-methoxyphenyl)-[1,1'-binaphthalene]-2,2'-diol 98%

Supplier: Ambeed
Warning

A641363-100MG A641363-1G A641363-250MG A641363-5G
77254-600EA 17 USD
77254-600 77254-602 77254-604 77254-606
(S)-3,3'-Bis(4-methoxyphenyl)-[1,1'-binaphthalene]-2,2'-diol 98%
(S)-3,3'-Bis(4-methoxyphenyl)-[1,1'-binaphthalene]-2,2'-diol

Precaución: for Research Use Only

Formula: C₃₄H₂₆O₄
MDL Number: MFCD30541640
CAS Number: 1199631-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR