6-Chloro-4-methoxy-1H-pyrazolo[3,4-d]pyrimidine 97%
Supplier: Ambeed
|
Warning
|
A641230-100MG
A641230-250MG
77267-060EA
30
USD
77267-060
77267-062
6-Chloro-4-methoxy-1H-pyrazolo[3,4-d]pyrimidine 97%
6-Chloro-4-methoxy-1H-pyrazolo[3,4-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₆H₅ClN₄O |
MDL Number:
MFCD24263253 CAS Number: 98138-75-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...